Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphB3 Antibody (115), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290645AF594
Description
EphB3 Monoclonal specifically detects EphB3 in Mouse samples. It is validated for ELISA.Specifications
EphB3 | |
Monoclonal | |
Alexa Fluor 594 | |
EC 2.7.10, EC 2.7.10.1, EK2, EPH receptor B3, EphB3, EPH-like kinase 2, EPH-like tyrosine kinase-2, ephrin type-B receptor 3, ETK2Hek2, HEK2, human embryo kinase 2, Tyro6, Tyrosine-protein kinase TYRO6 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EphB3 (NP_034273.1; Met 1-Thr 537). | |
0.1 mL | |
Protein Kinase | |
2049 | |
Mouse | |
Purified |
ELISA | |
115 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human LRPAP1 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction