Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphB3 Antibody (647354), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB56671MFV610
Description
EphB3 Monoclonal antibody specifically detects EphB3 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| EphB3 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 647354 | |
| Flow Cytometry, CyTOF-ready | |
| EC 2.7.10, EC 2.7.10.1, EK2, EPH receptor B3, EphB3, EPH-like kinase 2, EPH-like tyrosine kinase-2, ephrin type-B receptor 3, ETK2Hek2, HEK2, human embryo kinase 2, Tyro6, Tyrosine-protein kinase TYRO6 | |
| Mouse myeloma cell line NS0-derived recombinant human EphB3, Leu38-Ala550, Accession # P54753 | |
| 0.1 mL | |
| Protein Kinase | |
| 2049 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction