Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPHB3/Eph receptor B3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981231
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1231 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1231 Supplier Bioss Supplier No. BS11281R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EphB3, also known as Cek10, Hek2, Mdk5, Tyro6, and Sek4, is a member of the Eph receptor family, which binds members of the ephrin ligand family. The ephrin ligands and Eph receptors display reciprocal expression in vivo. Developing and adult neural tissue express nearly all of the Eph receptors and ephrin ligands. Ephs and ephrins play a significant role in angiogenesis.

Specifications

Antigen EPHB3/Eph receptor B3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EPHB3
Gene Accession No. P54753
Gene Alias AW456895; CAPB; Cek10; Developmental kinase 5; DRT; EK2; ELK; Embryonic kinase 2; EPH receptor B3; EPHB2; Ephb3; EPH-like kinase 2; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; Ephrin B1; Ephrin B2; Ephrin B3; Ephrin type-B receptor 3; EphrinB1; EphrinB2; EphrinB3; EPHT3; ERK; ETK2; HEK2; Hek5; HEK-6; human embryo kinase 2; Mdk5; mDK-5; MGC87492; NET; PCBC; Sek4; Tyro5; TYRO6; Tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO6
Gene Symbols EPHB3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EPHB3/Eph receptor B3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2049
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.