Learn More
Invitrogen™ EphB3 Polyclonal Antibody

Description
Immunogen sequence: LSRFLEDDPS DPTYTSSLGG KIPIRWTAPE AIAYRKFTSA SDVWSYGIVM WEVMSYGERP YWDMSNQDVI NAVEQDYRLP PPMDCPTALH QLMLDCWVRD RNLRPKFSQI VNTLDKLIRN AASLKVIASA QSGMSQPLLD RTVPDYTTFT TVGDWLDAIK MGRYKESFVS AGFASFDLVA QMTAEDLLRI GVTLAGHQKK ILSSIQDMRL QMNQTLPVQV Positive Samples: HeLa; Cellular Location: Cell membrane, Cell projection, Single-pass type I membrane protein, dendrite.
Specifications
Specifications
| Antigen | EphB3 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 4.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | EPHB3 |
| Gene Accession No. | P54753, P54754 |
| Gene Alias | AW456895; Cek10; Developmental kinase 5; EK2; Embryonic kinase 2; EPH receptor B3; EPHB3; EPH-like kinase 2; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; Ephrin type-B receptor 3; Etk2; HEK2; human embryo kinase 2; Mdk5; mDK-5; Sek4; TYRO6; tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO6 |
| Gene Symbols | EPHB3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.