Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EphB3 Recombinant Rabbit Monoclonal Antibody (001)
GREENER_CHOICE

Catalog No. PIMA536048
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA536048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA536048 Supplier Invitrogen™ Supplier No. MA536048
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

EphB3 Recombinant Monoclonal Antibody for Flow

EPHB3 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. The ephrin ligands and Eph receptors display reciprocal expression in vivo. Developing and adult neural tissue express nearly all of the Eph receptors and ephrin ligands. Ephs and ephrins play a significant role in angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphB3
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 001
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene EPHB3
Gene Accession No. P54754
Gene Alias AW456895; Cek10; Developmental kinase 5; EK2; Embryonic kinase 2; EPH receptor B3; EPHB3; EPH-like kinase 2; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; Ephrin type-B receptor 3; Etk2; HEK2; human embryo kinase 2; Mdk5; mDK-5; Sek4; TYRO6; tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO6
Gene Symbols EPHB3
Host Species Rabbit
Immunogen Recombinant Mouse EphB3/HEK2/Eph Receptor B3 Protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13845
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.