Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EphB3 Recombinant Rabbit Monoclonal Antibody (JE54-73)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA536195
Description
EphB3 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
EPHB3 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. The ephrin ligands and Eph receptors display reciprocal expression in vivo. Developing and adult neural tissue express nearly all of the Eph receptors and ephrin ligands. Ephs and ephrins play a significant role in angiogenesis.
Specifications
EphB3 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P54753 | |
EPHB3 | |
Recombinant protein within Human Eph receptor B3 aa 80-220. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
JE54-73 | |
Unconjugated | |
EPHB3 | |
AW456895; Cek10; Developmental kinase 5; EK2; Embryonic kinase 2; EPH receptor B3; EPHB3; EPH-like kinase 2; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; Ephrin type-B receptor 3; Etk2; HEK2; human embryo kinase 2; Mdk5; mDK-5; Sek4; TYRO6; tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO6 | |
Rabbit | |
Protein A | |
RUO | |
2049, 287989 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction