Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphB4 Antibody (047), Alexa Fluor™ 700 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289353AF700
Description
EphB4 Monoclonal antibody specifically detects EphB4 in Human samples. It is validated for ELISASpecifications
EphB4 | |
Monoclonal | |
Alexa Fluor 700 | |
EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB4 (NP_004435.3; Met1-Ala539). | |
0.1 mL | |
Protein Kinase | |
2050 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
047 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction