Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphB4 Rabbit anti-Human, Clone: H1, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | EphB4 |
---|---|
Clone | H1 |
Dilution | ELISA 1:1000-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
EphB4 Monoclonal specifically detects EphB4 in Human samples. It is validated for ELISA.Specifications
EphB4 | |
ELISA 1:1000-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.10, EC 2.7.10.1, EPH receptor B4, EphB4, ephrin type-B receptor 4, hepatoma transmembrane kinase, HTKephrin receptor EphB4, MYK1, soluble EPHB4 variant 1, soluble EPHB4 variant 2, soluble EPHB4 variant 3, Tyro11, Tyrosine-protein kinase receptor HTK, Tyrosine-protein kinase TYRO11 | |
Recombinant Human EphB4 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
H1 | |
ELISA | |
Unconjugated | |
Rabbit | |
Protein Kinase | |
0.2 um filtered solution in PBS | |
2050 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title