Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EphB6 Antibody (037), HRP , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289328H
Description
EphB6 Monoclonal antibody specifically detects EphB6 in Human samples. It is validated for ELISASpecifications
EphB6 | |
Monoclonal | |
HRP | |
EC 2.7.10.1, EPH receptor B6, EphB6, ephrin type-B receptor 6, HEPTyrosine-protein kinase-defective receptor EPH-6, MGC129910, MGC129911 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB6 (NP_004436.1; Met 1-Ser 579). | |
0.1 mL | |
Protein Kinase | |
2051 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
037 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction