Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephexin-1/EPHEXIN Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981382
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1382 Supplier Bioss Supplier No. BS11458R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acts as a guanine nucleotide exchange factor (GEF) which differentially activates the GTPases RHOA, RAC1 and CDC42. Plays a role in axon guidance regulating ephrin-induced growth cone collapse and dendritic spine morphogenesis. Upon activation by ephrin through EPHA4, the GEF activity switches toward RHOA resulting in its activation. Activated RHOA promotes cone retraction at the expense of RAC1- and CDC42-stimulated growth cone extension.

Specifications

Antigen Ephexin-1/EPHEXIN
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NGEF
Gene Accession No. Q8N5V2
Gene Alias ARHGEF27; Besh3; brain-enriched SH3-domain protein Besh3; ephexin; Ephexin-1; Eph-interacting exchange protein; Neuronal guanine nucleotide exchange factor; NGEF; Tims2; transforming immortalised mammary epithelial sequence 2
Gene Symbols NGEF
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EPHEXIN .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25791
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.