Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ephrin-A1 Antibody (067), Alexa Fluor™ 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289659AF350
Description
Ephrin-A1 Monoclonal antibody specifically detects Ephrin-A1 in Rat samples. It is validated for ELISASpecifications
Ephrin-A1 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Rat | |
Purified |
ELISA | |
067 | |
ELISA | |
ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rat Ephrin-A1 (P97553; Met1-His181). | |
0.1 mL | |
Cancer | |
1942 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction