Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ephrin-A1 Antibody (114), Janelia Fluor™ 549, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289691JF549
Description
Ephrin-A1 Monoclonal specifically detects Ephrin-A1 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
Ephrin-A1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human EphrinA5/ EFNA5 Human EphrinA3/ EFNA3 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
114 | |
ELISA, Sandwich ELISA Detection | |
ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Ephrin-A1 (NP_004419.2; Met 1-Ser 182). | |
0.1 mL | |
Cancer | |
1942 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction