Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin-A1 Antibody (RM0224-6D31), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB212277405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB212277405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB212277405 Supplier Novus Biologicals Supplier No. NBP212277AF405
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ephrin-A1 Monoclonal specifically detects Ephrin-A1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Ephrin-A1
Applications Western Blot
Classification Monoclonal
Clone RM0224-6D31
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4
Gene Symbols EFNA1
Host Species Rat
Immunogen Mouse recombinant Ephrin-A1
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1942
Test Specificity Detects mouse Ephrin-A1 with WB and shows weaker cross-reactivity with mouse Ephrin-A2.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.