Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ephrin-A1 Antibody (RM0224-6D31), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212277B
Description
Ephrin-A1 Monoclonal specifically detects Ephrin-A1 in Mouse samples. It is validated for Western Blot.Specifications
Ephrin-A1 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
EFNA1 | |
Mouse recombinant Ephrin-A1 | |
0.1 mL | |
Primary | |
Detects mouse Ephrin-A1 with WB and shows weaker cross-reactivity with mouse Ephrin-A2. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0224-6D31 | |
Western Blot | |
ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1, EPLG1TNF alpha-induced protein 4, Immediate early response protein B61, LERK1LERK-1, ligand of eph-related kinase 1, TNFAIP4B61, Tumor necrosis factor alpha-induced protein 4, tumor necrosis factor, alpha-induced protein 4 | |
Rat | |
Protein A or G purified | |
Cancer | |
1942 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction