Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ephrin A1 Monoclonal Antibody (3F7H7F6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA543006
This item is not returnable.
View return policy
Description
Specificity: Human EphrinA1/EFNA1
This gene encodes a member of the ephrin family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. This gene encodes an EFNA class ephrin which binds to the EPHA2, EPHA4, EPHA5, EPHA6, and EPHA7 receptors. Two transcript variants that encode different isoforms were identified through sequence analysis.
Specifications
Ephrin A1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P20827 | |
EFNA1 | |
Recombinant Human EFNA1 protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
3F7H7F6 | |
Unconjugated | |
EFNA1 | |
AI325262; B61; ECKLG; EFL1; Efna1; Epgl1; EPH-related receptor tyrosine kinase ligand 1; ephrin A1; ephrin-A1; Ephrin-A1, secreted form; Epl1; EPLG1; Immediate early response protein B61; Lerk1; LERK-1; ligand of eph-related kinase 1; TNF alpha-induced protein 4; TNFAIP4; tumor necrosis factor alpha-induced protein 4; tumor necrosis factor, alpha-induced protein 4 | |
Mouse | |
Protein A | |
RUO | |
1942 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction