Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ephrin A1 Monoclonal Antibody (A7B10)
GREENER_CHOICE

Catalog No. FEMA550588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550588 Supplier Invitrogen™ Supplier No. MA550588
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: SK-Br-3 cell lysates, SK-Br-3. Predicted band size: 24 kDa Subcellular Location: Cell membrane, Secreted.

This gene encodes a member of the ephrin family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. This gene encodes an EFNA class ephrin which binds to the EPHA2, EPHA4, EPHA5, EPHA6, and EPHA7 receptors. Two transcript variants that encode different isoforms were identified through sequence analysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin A1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A7B10
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene EFNA1
Gene Accession No. P20827
Gene Alias AI325262; B61; ECKLG; EFL1; Efna1; Epgl1; EPH-related receptor tyrosine kinase ligand 1; ephrin A1; ephrin-A1; Ephrin-A1, secreted form; Epl1; EPLG1; Immediate early response protein B61; Lerk1; LERK-1; ligand of eph-related kinase 1; TNF alpha-induced protein 4; TNFAIP4; tumor necrosis factor alpha-induced protein 4; tumor necrosis factor, alpha-induced protein 4
Gene Symbols EFNA1
Host Species Mouse
Immunogen Recombinant protein within Human Ephrin A1 aa 13-190.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1942
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.