Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ephrin A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519397
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519397 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519397 Supplier Invitrogen™ Supplier No. PA519397
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, human and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

This gene encodes a member of the ephrin family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. This gene encodes an EFNA class ephrin which binds to the EPHA2, EPHA4, EPHA5, EPHA6, and EPHA7 receptors. Two transcript variants that encode different isoforms were identified through sequence analysis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin A1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene EFNA1
Gene Accession No. P20827, P52793, P97553
Gene Alias AI325262; B61; ECKLG; EFL1; Efna1; Epgl1; EPH-related receptor tyrosine kinase ligand 1; ephrin A1; ephrin-A1; Ephrin-A1, secreted form; Epl1; EPLG1; Immediate early response protein B61; Lerk1; LERK-1; ligand of eph-related kinase 1; TNF alpha-induced protein 4; TNFAIP4; tumor necrosis factor alpha-induced protein 4; tumor necrosis factor, alpha-induced protein 4
Gene Symbols EFNA1
Host Species Goat
Immunogen Synthetic peptide sequence (KVTVSGKITHSP) corresponding to the internal amino acids of EFNA1 (aa 144-155).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13636, 1942, 94268
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.