Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ephrin-A2 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298103JF646
Description
Ephrin-A2 Polyclonal antibody specifically detects Ephrin-A2 in Mouse samples. It is validated for ELISASpecifications
Ephrin-A2 | |
Polyclonal | |
ELISA | |
ELF-1, EPH-related receptor tyrosine kinase ligand 6, ephrin-A2, EPLG6HEK7 ligand, HEK7-L, HEK7-ligand, LERK6LERK-6, ligand of eph-related kinase 6 | |
Rabbits immunized with purified, recombinant Mouse Ephrin-A2 (Accession#: NP_031935.3, Met 1-Asn 184) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
1943 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction