Learn More
Invitrogen™ Ephrin A2 Monoclonal Antibody (OTI2B7)

Description
Ephrin A2 Monoclonal Antibody for ICC/IF, Western Blot
Specifications
Specifications
| Antigen | Ephrin A2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2B7 |
| Concentration | 1.19 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | EFNA2 |
| Gene Accession No. | O43921 |
| Gene Alias | CEK7L; CEK7-L; CEK7-ligand; EFNA2; Elf1; ELF-1; EPH related receptor tyrosine kinase ligand 6; eph-related receptor tyrosine kinase ligand 6; ephrin A2; ephrin A-2; ephrin A6; EphrinA2; ephrin-A2; Epl6; EPLG6; HEK7 ligand; HEK7-L; Lerk6; LERK-6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.