Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ephrin A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111624
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111624 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111624 Supplier Invitrogen™ Supplier No. PA5111624
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGHEYYYISA TPPNAVDRPC LRLKVYVRPT NETLYEAPEP IFTSNNSCSS PGGCRLFLST IPVLWTLLG Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 88%.

The Eph subfamily represents the largest group of receptor protein tyrosine kinases identified to date. While the biological activities of these receptors have yet to be determined, there is increasing evidence that they are involved in central nervous system function and in development. The Eph subfamily receptors of human origin (and their murine/avian homologs) include EphA1(Eph), EphA2 (Eck), EphA3 (Hek4), EphA4 (Hek8), EphA5 (Hek7), EphA6 (Hek12), EphA7 (Hek11/MDK1), EphA8 (Hek3), EphB1 (Hek6), EphB2 (Hek5), EphB3(Cek10, Hek2), EphB4 (Htk), EphB5 (Hek9) and EphB6 (Mep). Ligands for Eph receptors include ephrin-A4 (LERK-4) which binds EphA3 and EphB1. In addition, Ephrin-A2 (Elf-1) has been described as the ligand for EphA4, ephrin-A3 (Ehk1-L) as the ligand for EphA5 and ephrin-B2 (Htk-L) as the ligand for EphB4 (Htk).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin A2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFNA2
Gene Accession No. O43921
Gene Alias CEK7L; CEK7-L; CEK7-ligand; EFNA2; Elf1; ELF-1; EPH related receptor tyrosine kinase ligand 6; eph-related receptor tyrosine kinase ligand 6; ephrin A2; ephrin A-2; ephrin A6; EphrinA2; ephrin-A2; Epl6; EPLG6; HEK7 ligand; HEK7-L; Lerk6; LERK-6
Gene Symbols EFNA2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Ephrin A2. Recombinant protein control fragment (Product #RP-108532).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1943
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.