Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ephrin-A3 Antibody (MM0262-7N12) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $560.50
Specifications
Antigen | Ephrin-A3 |
---|---|
Clone | MM0262-7N12 |
Concentration | LYOPH |
Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin 15 ug/ml |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Description
Ephrin-A3 Monoclonal specifically detects Ephrin-A3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Ephrin-A3 | |
LYOPH | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Growth and Development, Neuronal Cell Markers | |
AAA52368 | |
1944 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
MM0262-7N12 | |
Western Blot 1:500 - 1:1000, Immunohistochemistry-Paraffin 15 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EFL2, EFL-2, EHK1-L, EPH-related receptor tyrosine kinase ligand 3, ephrin-A3, EPLG3EHK1 ligand, LERK3LERK-3, ligand of eph-related kinase 3 | |
EFNA3 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title