Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ephrin-A3 Antibody (OTI4A4), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270623V
Description
Ephrin-A3 Monoclonal specifically detects Ephrin-A3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
Ephrin-A3 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EFNA3 | |
Recombinant protein fragment corresponding to amino acids 23-214 of human Ephrin-A3 (NP_004943) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Immunohistochemistry | |
OTI4A4 | |
Immunohistochemistry | |
EFL2, EFL-2, EHK1-L, EPH-related receptor tyrosine kinase ligand 3, ephrin-A3, EPLG3EHK1 ligand, LERK3LERK-3, ligand of eph-related kinase 3 | |
Mouse | |
Affinity Purified | |
Growth and Development, Neuronal Cell Markers | |
1944.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction