Learn More
Ephrin-A4 Antibody (MM0263-9H13), Alexa Fluor™ 594, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP212280AF594
Catalog No. NP212280A94
Only null left
Add to Cart
Description
Description
Ephrin-A4 Monoclonal specifically detects Ephrin-A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Ephrin-A4
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
EFNA4
Human recombinant Ephrin-A4 (extracellular domain)
Primary
Human
IgG2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
MM0263-9H13
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
EFL4, EPH-related receptor tyrosine kinase ligand 4, ephrin-A4, EPLG4MGC125826, LERK-4, LERK4FLJ57652, ligand of eph-related kinase 4
Mouse
0.1 mL
1945
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.