Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EphA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA130293
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA130293 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA130293 Supplier Invitrogen™ Supplier No. PA130293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NCI-H460 cell lysate. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

EPHA4 belongs to the ephrin receptor subfamily of the protein tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediated developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA4 encodes a protein that binds ephrin-A ligands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EphA4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene EPHA4
Gene Accession No. P54764, Q03137
Gene Alias 2900005C20Rik; AI385584; Cek8; Cek8 protein; EK8; epa4; EPH receptor A4; Epha4; EPH-like kinase 8; Ephrin type-A receptor 4; Hek8; rb; receptor protein-tyrosine kinase HEK8; RGD1560587; Sek; Sek1; Tyro1; TYRO1 protein tyrosine kinase; tyrosine-protein kinase receptor MPK-3; Tyrosine-protein kinase receptor SEK; tyrosine-protein kinase receptor SEK-1; Tyrosine-protein kinase TYRO1
Gene Symbols EPHA4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 875-904 amino acids from the C-terminal region of human EphA4.
Purification Method Ammonium sulfate precipitation
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13838, 2043
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.