Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ephrin-B1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Ephrin-B |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
Ephrin-B1 Polyclonal specifically detects Ephrin-B1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Ephrin-B | |
Polyclonal | |
Rabbit | |
Cellular Signaling, Neuroscience | |
EFL3, EFL-3, ELK ligand, Elk-L, ephrin-B1, LERK-2 | |
EFNB1 | |
IgG | |
Affinity Purified |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
1947 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VGQDPNAVTPEQLTTSRPSKEADNTVKMATQAPGSRGSLGDSDGKHETVNQEEKSGPGA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title