Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin B1, B2, B3 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992340
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2340 100 μg
50-199-2341 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2340 Supplier Rockland Immunochemicals Supplier No. 600401MP0
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This affinity purified antibody is directed against human Ephrin B1, B2, B3. A BLAST analysis was used to suggest cross-reactivity with the antigen based on 100% homology with Ephrin-B1 to human, mouse, rat, and Gallus gallus; 100% homology with Ephrin-B2 to mouse; 100% homology with Ephrin-B3 to human and mouse.

Eph receptors and ephrins exhibit complementary expression in many tissues during embryogenesis indicating that bidirectional activation of Eph receptors and ephrin-B proteins may occur at expression domain interfaces. Ephrin-B1 transduces outside-in signals through C-terminal protein interactions that effect integrin-mediated cell attachment and migration. The distribution of ephrin-B1 in the developing retina suggests that it influences retinal axon mapping along the dorsal-ventral axis and may be involved in intratectal development The transmembrane ligand ephrin-B2 and its receptor tyrosine kinase EphB4 are specifically expressed on arterial and venous endothelial cells, respectively.

Specifications

Antigen Ephrin B1, B2, B3
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene EFNB1
Gene Accession No. O35393, P52795, P52799, P52800, P98172, Q15768
Gene Alias CEK5 receptor ligand; CEK5-L; CFND; CFNS; EFB1; EFL3; EFL-3; EFL6; EFL-6; Efnb1; Efnb2; Efnb3; Elf2; ELF-2; ELF-3; ELK ligand; ELK-L; Elk-L3; EPH-related receptor transmembrane ligand ELK-L3; EPH-related receptor tyrosine kinase ligand 2; EPH-related receptor tyrosine kinase ligand 5; EPH-related receptor tyrosine kinase ligand 8; ephrin B1; ephrin B2; ephrin B3; ephrin ligand B3; Ephrin-B1; Ephrin-B1 C-terminal fragment; Ephrin-B1 CTF; Ephrin-B1 ICD; Ephrin-B1 intracellular domain; Ephrin-B2; Ephrin-B3; Epl2; Epl5; Epl8; Eplg2; EPLG5; EPLG8; HTK ligand; HTKL; HTK-L; Lerk2; LERK-2; Lerk5; LERK-5; LERK8; LERK-8; ligand of eph-related kinase 5; NLERK-1; NLERK-2; Stimulated by retinoic acid gene 1 protein; Stra1
Gene Symbols EFNB1, EFNB2, EFNB3
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13641, 13642, 13643, 1947, 1948, 1949, 779011
Target Species Human, Mouse, X. laevis
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.