Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ephrin-B1, p Tyr298 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300274
Description
Ephrin-B1 Polyclonal specifically detects Ephrin-B1 in Human, Rat samples. It is validated for Western Blot.Specifications
Ephrin-B1 (p Tyr298) | |
Polyclonal | |
Western Blot 1:1000 | |
F7B9D9, F7B9D9, F7B9D9, F7B9D9, F7B9D9, F7B9D9, F7B9D9 | |
EFNB1 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ephrin-B1 conjugated to KLH. Accession # P28693 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼46k EphrinB protein phosphorylated at Tyr298. Immunolabeling of the EphrinB band is blocked by lambda-phosphatase treatment. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
EFL3, EFL-3, ELK ligand, Elk-L, ephrin-B1, LERK-2 | |
Rabbit | |
46 kDa | |
0.1 mL | |
Cellular Signaling, Neuroscience | |
1947 | |
Human, Rat, Xenopus species | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction