Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ephrin-B2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298424MFV610
Description
Ephrin-B2 Polyclonal antibody specifically detects Ephrin-B2 in Rat samples. It is validated for ELISASpecifications
Ephrin-B2 | |
Polyclonal | |
ELISA | |
ephrin B2, EPLG5, Htk-L, HTKL, LERK5 | |
Produced in rabbits immunized with purified, recombinant Rat Ephrin-B2 (Accession#: NP_001100798.1; Met1-Ala229). | |
0.1 mL | |
Neuroscience, Signal Transduction | |
1948 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction