Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ephrin B2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588892
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588892 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588892 Supplier Invitrogen™ Supplier No. PA588892
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IVLEPIYWNS SNSKFLPGQG LVLYPQIGDK LDIICPKVDS KTVGQYEYYK VYMVDKDQAD RCTIKKENTP LLNCAKPDQD IKFTIKFQEF SPNLWGLEFQ KNKDYYIIST SNGSLEGLDN QEGGVCQTRA MKILMKVGQD ASSAGSTRNK DPTRRPELEA GTNGRSSTTS PFVKPNPGSS TDGNSAGHSG NNILGSEVAL FA; Positive Samples: Mouse brain, Rat brain; Cellular Location: Membrane, Single-pass type I membrane protein.

This gene encodes a member of the ephrin (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. This gene encodes an EFNB class ephrin which binds to the EPHB4 and EPHA3 receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin B2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Efnb2
Gene Accession No. P52800
Gene Alias EFNB2; efnb2a; Elf2; ELF-2; EPH-related receptor tyrosine kinase ligand 5; ephrin B2; ephrin B2a; ephrin-B2; Ephrin-B2a; Epl5; EPLG5; HTK ligand; Htkl; HTK-L; id:ibd5072; Lerk5; LERK-5; ligand of eph-related kinase 5; MGC126226; MGC126227; MGC126228; NLERK-1
Gene Symbols Efnb2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 28-229 of human Ephrin B2 (NP_0040841).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13642, 306636
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.