Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ephrin-B3 Antibody (MM0264- 12X1), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212283MFV500
Description
Ephrin-B3 Monoclonal antibody specifically detects Ephrin-B3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Ephrin-B3 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MM0264- 12X1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EPH-related receptor tyrosine kinase ligand 8, Ephrin B3, ephrin-B3, EPLG8EPH-related receptor transmembrane ligand ELK-L3, LERK-8, LERK8EFL6 | |
Human recombinant Ephrin-B3 (extracellular domain) | |
0.1 mL | |
Cardiovascular Biology, Neuroscience | |
1949 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction