Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ephrin B3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110638 Supplier Invitrogen™ Supplier No. PA5110638
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSSPNYEFYK LYLVGGAQGR RCEAPPAPNL LLTCDRPDLD LRFTIKFQEY SPNLWGHEFR SHHDYYIIAT SDGTREGLES LQGGVCLTRG MKVLLRVGQS PRGGAVPRKP VSEMPMERDR GAAHSLEPGK ENLPGDPTSN ATS Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 96%.

EFNB3, a member of the ephrin gene family, is important in brain development as well as in its maintenance. Moreover, since levels of EFNB3 expression were particularly high in several forebrain subregions compared to other brain subregions, it may play a pivotal role in forebrain function. The EPH and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, particularly in the nervous system. EPH Receptors typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin ligands and receptors have been named by the Eph Nomenclature Committee (1997). Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are similarly divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ephrin B3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFNB3
Gene Accession No. Q15768
Gene Alias EFL6; EFL-6; Efnb3; ELF-3; Elk-L3; EPH-related receptor transmembrane ligand ELK-L3; eph-related receptor tyrosine kinase ligand 8; ephrin B3; ephrin ligand B3; ephrin-B3; Epl8; EPLG8; LERK8; LERK-8; NLERK-2
Gene Symbols EFNB3
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Ephrin B3. Recombinant protein control fragment (Product #RP-94752).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1949
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.