Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ephrin Receptor A8 Rabbit anti-Human, Mouse, Polyclonal, FabGennix

Catalog No. 502206928
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6928 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6928 Supplier Fabgennix Inc Supplier No. EPHAR8801AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ephrin Receptor A8 Polyclonal antibody specifically detects Ephrin Receptor A8 in Human, Mouse samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Ephrin Receptor A8
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Epha8
Gene Accession No. O09127, P29322
Gene Alias AW047546; Eek; EK3; epa8; EPH- and ELK-related kinase; EPH- and ELK-related tyrosine kinase; Eph receptor A8; Epha8; EPH-like kinase 3; Ephrin type-A receptor 8; Ephrin type-A receptor 8 precursor (Tyrosine-protein kinase receptor EEK) (EPH-and ELK-related kinase); Hek3; hydroxyaryl-protein kinase; KIAA1459; mKIAA1459; protein-tyrosine kinase; tyrosine-protein kinase receptor EEK; tyrosylprotein kinase
Host Species Rabbit
Immunogen Synthetic peptide corresponding to unique amino acid sequences on human EphA R8 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13842, 2046
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.