Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPHX1 Antibody (2E5), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP130163AF350
Description
EPHX1 Monoclonal specifically detects EPHX1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
EPHX1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
EPHX1 | |
Recombinant human EPHX1 (21-455aa) purified from E. coli | |
Primary | |
Human | |
IgG2a κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
200000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 3.3.2.9, EPHX, EPOX, Epoxide hydratase, epoxide hydrolase 1, epoxide hydrolase 1, microsomal (xenobiotic), MEH, Microsomal epoxide hydrolase | |
Mouse | |
0.1 mL | |
2052 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction