Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPHX1 Mouse anti-Human, Clone: 15B2AD9, Abnova™

Catalog No. 89035447 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-447 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-447 Supplier Abnova Corporation Supplier No. MAB14042

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against human EPHX1.

  • High specificity
  • High reactivity

Specifications

Antigen EPHX1
Applications ELISA, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 15B2AD9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against human EPHX1.
Dilution ELISA Immunocytochemistry (4 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Formulation In HEPES (0.02% sodium azide).
Gene Accession No. P07099
Gene Alias EPHX/EPOX/MEH
Gene Symbols EPHX1
Host Species Mouse
Immunogen Human EPHX1.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2052
Target Species Human
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.