Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPHX1 Mouse anti-Human, Clone: OTI3A10, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA600437

Description
Biotransformation enzyme that catalyzes the hydrolysis of arene and aliphatic epoxides to less reactive and more water soluble dihydrodiols by the trans addition of water. May play a role in the metabolism of endogenous lipids such as epoxide-containing fatty acids.Specifications
EPHX1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with <5% trehalose and 0.05% sodium azide | |
P07099 | |
EPHX1 | |
Full length human recombit protein of human EPHX1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
OTI3A10 | |
Unconjugated | |
EPHX1 | |
EPHX, EPOX, HYL1, MEH | |
Mouse | |
Affinity Chromatography | |
RUO | |
2052 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction