Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPHX1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522082

Catalog No. PIPA522082


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, H1299, HeLa. Predicted reactivity: Mouse (87%), Rat (87%), Pig (90%), Rabbit (88%), Rhesus Monkey (98%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Biotransformation enzyme that catalyzes the hydrolysis of arene and aliphatic epoxides to less reactive and more water soluble dihydrodiols by the trans addition of water. May play a role in the metabolism of endogenous lipids such as epoxide-containing fatty acids.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

EPHX1
Polyclonal
Unconjugated
EPHX1
AI195553; Eph1; Eph-1; EPHX; Ephx1; EPOX; Epoxide hydratase; epoxide hydrolase 1; epoxide hydrolase 1 (microsomal xenobiotic hydrolase); epoxide hydrolase 1 microsomal; epoxide hydrolase 1, microsomal; epoxide hydrolase 1, microsomal (xenobiotic); Epxh1; HYL1; liver microsomal xenobiotic epoxide hydrolase; mEH; MEH8; microsomal epoxide hydrolase; microsomal epoxide hydrolase (EC 3.3.2.3)
Rabbit
Antigen affinity chromatography
RUO
2052
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P07099
EPHX1
Recombinant protein encompassing a sequence within the center region of human EPHX1. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.