Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPHX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582730 Supplier Invitrogen™ Supplier No. PA582730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DKEETLPLED GWWGPGTRSA AREDDSIRPF KVETSDEEIH DLHQRIDKFR FTPPLEDSCF HYGFNSNYLK KVISYWRNEF DWKKQVEILN RHPHFKTKIE GLDIHFI Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 83%.

Biotransformation enzyme that catalyzes the hydrolysis of arene and aliphatic epoxides to less reactive and more water soluble dihydrodiols by the trans addition of water. May play a role in the metabolism of endogenous lipids such as epoxide-containing fatty acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPHX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EPHX1
Gene Accession No. P07099
Gene Alias AI195553; Eph1; Eph-1; EPHX; Ephx1; EPOX; Epoxide hydratase; epoxide hydrolase 1; epoxide hydrolase 1 (microsomal xenobiotic hydrolase); epoxide hydrolase 1 microsomal; epoxide hydrolase 1, microsomal; epoxide hydrolase 1, microsomal (xenobiotic); Epxh1; HYL1; liver microsomal xenobiotic epoxide hydrolase; mEH; MEH8; microsomal epoxide hydrolase; microsomal epoxide hydrolase (EC 3.3.2.3)
Gene Symbols EPHX1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EPHX1. Recombinant protein control fragment (Product #RP-92515).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2052
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.