Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EPHX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583932
Description
Immunogen sequence: LPAGHTPKPL LMVHGWPGSF YEFYKIIPLL TDPKNHGLSD EHVFEVICPS IPGYGFSEAS SKKGFNSVAT ARIFYKLMLR LGFQEFYIQG GDWGSLICTN.
Biotransformation enzyme that catalyzes the hydrolysis of arene and aliphatic epoxides to less reactive and more water soluble dihydrodiols by the trans addition of water. May play a role in the metabolism of endogenous lipids such as epoxide-containing fatty acids.
Specifications
EPHX1 | |
Polyclonal | |
Unconjugated | |
EPHX1 | |
AI195553; Eph1; Eph-1; EPHX; Ephx1; EPOX; Epoxide hydratase; epoxide hydrolase 1; epoxide hydrolase 1 (microsomal xenobiotic hydrolase); epoxide hydrolase 1 microsomal; epoxide hydrolase 1, microsomal; epoxide hydrolase 1, microsomal (xenobiotic); Epxh1; HYL1; liver microsomal xenobiotic epoxide hydrolase; mEH; MEH8; microsomal epoxide hydrolase; microsomal epoxide hydrolase (EC 3.3.2.3) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2052 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P07099 | |
EPHX1 | |
Recombinant protein corresponding to Human EPHX1. Recombinant protein control fragment (Product #RP-103038). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction