Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EPHX2 Antibody (OTI1A6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270648H
Description
EPHX2 Monoclonal specifically detects EPHX2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
EPHX2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
EPHX2 | |
Full length recombinant protein of human EPHX2 (NP_001970) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1A6 | |
Western Blot, Immunohistochemistry | |
CEH, Cytosolic epoxide hydrolase, EC 3.3.2.10, Epoxide hydratase, epoxide hydrolase 2, epoxide hydrolase 2, cytoplasmic, epoxide hydrolase 2, cytosolic, epoxide hydrolase, soluble, SEH, Soluble epoxide hydrolase | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
2053.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction