Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EPHX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532755
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532755 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532755 Supplier Invitrogen™ Supplier No. PA532755
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Elephant (100%); Bat, Bovine, Horse (94%); Dog, Rabbit, Pig (88%); Mouse, Rat, Hamster, Stickleback (82%). Immunohistochemistry with formalin-fixed paraffin-embedded tissues requires pretreatment with Proteinase K.

This gene encodes a member of the epoxide hydrolase family. The protein, found in both the cytosol and peroxisomes, binds to specific epoxides and converts them to the corresponding dihydrodiols. Mutations in this gene have been associated with familial hypercholesterolemia. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPHX2
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene EPHX2
Gene Accession No. P34913
Gene Alias bifunctional epoxide hydrolase 2; CEH; cytosolic epoxide hydrolase; Cytosolic epoxide hydrolase 2; Eph2; Ephx2; Epoxide hydratase; epoxide hydrolase 2; epoxide hydrolase 2, cytoplasmic; epoxide hydrolase 2, cytosolic; epoxide hydrolase 2C; epoxide hydrolase, soluble; Lipid-phosphate phosphatase; SEH; sEP; Soluble epoxide hydrolase
Gene Symbols EPHX2
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from C-Terminus of human EPHX2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2053
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.