Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Epiandrosterone Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175128
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175128 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175128 Supplier Invitrogen™ Supplier No. PA175128
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 1.25 μg/mL. Sensitivity: 10 ng/mL Epiandrosterone produces 74% inhibition in a competitive ELISA employing Epiandrosterone polyclonal antibody.

Epiandrosterone is a dehydroepiandrosterone metabolite and a precursor of testosterone and estradiol with hypolipidemic and anabolic property. Epiandrosterone, a potential neurosteroid, appears to bind to the gamma-aminobutyric acid (GABA)/benzodiazepine-receptor complex (GABA-RC), acting as a negative non-competitive modulator of GABA-RC as well as signal through the N-methyl-D-aspartate receptor. In addition this agent inhibits the pentose phosphate pathway (PPP) thereby dilating blood vessels pre-contracted by partial depolarization. Also, epiandrosterone inhibits the synthesis of thromboxane A2 in activated platelets, reduces plasma plasminogen activator inhibitor type 1 and tissue plasminogen activator antigen, increases serum levels of insulin-like growth factor 1 and increases cyclic guanosine monophosphate and nitric oxide synthesis. These effects may improve circulation in the microvasculature. [National Center for Biotechnology Information. PubChem Compound Database; CID=441302].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Epiandrosterone
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias 3beta-androsterone; 3Beta-hydroxy-5alpha-androstan-17-one; 5alpha-androstan-3beta-ol-17-one; C19H30O2; Epiandrosterone; Isoandrosterone; Trans-Androsterone
Host Species Sheep
Immunogen Epiandrosterone(17)-BTG.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.