Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Epicatechin Gallate, PROMO, Thermo Scientific Chemicals

Catalog No. J60814MAPM
Click to view available options
Quantity:
10 mg

A natural product from green tea that induces apoptosis

Applications

(-)-Epicatechin gallate is used as an antioxidant investigated as an adjunct treatment of some methyicillin resistant bacteria. Epicatechin gallate binds to the cell membrane in methicillin-resistant Staphylococcus aureus which decreases the fluidity of the bilayer and induces changes in gene expression. One of the catechin isomers and a potent antioxidant that can modulate a wide range of membrane proteins. Its bilayer-modifying potency was tested using gramicidin A (gA) channels as probes. All the catechins alter gA channel function and modify bilayer properties, with a 500-fold range in potency.

Solubility

Soluble in water, acetone, DMSO, methanol.

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 to 8°C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257-08-5
Molecular Formula C22H18O10
Molecular Weight (g/mol) 442.38 g/mol
MDL Number MFCD00075936
InChI Key LSHVYAFMTMFKBA-TZIWHRDSSA-N
Synonym =--epicatechin gallate, epicatechin gallate, =--epicatechin-3-o-gallate, l-epicatechin gallate, =--epicatechin-3-gallate, =--epicatechingallate, epicatechin monogallate, --epicatechin 3-o-gallate, teatannin, epicatechol, gallate
PubChem CID 107905
ChEBI CHEBI:70255
IUPAC Name [(2R,3R)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3,4-dihydro-2H-chromen-3-yl] 3,4,5-trihydroxybenzoate
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)OC(=O)C4=CC(=C(C(=C4)O)O)O

Specifications

Color White
Quantity 10 mg
Solubility Information Soluble in water, acetone, DMSO, methanol.
Physical Form Powder
Chemical Name or Material (-)-Epicatechin gallate
RTECSNumber DH9030000
TSCA No
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.