Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epichlorohydrin, 99%

Catalog No. AC117780025
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117780025 2.5 L Glass bottle
AC117780250 25 mL Glass bottle
AC117780010 1 L Glass bottle
AC117780100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC117780025 Supplier Thermo Scientific Chemicals Supplier No. 117780025
Only null left
Add to Cart

CAS: 106-89-8 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-89-8
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
InChI Key BRLQWZUYTZBJKN-UHFFFAOYSA-N
Synonym epichlorohydrin, 2-chloromethyl oxirane, epichlorhydrin, 1-chloro-2,3-epoxypropane, glycidyl chloride, oxirane, chloromethyl, epichlorhydrine, 1,2-epoxy-3-chloropropane, 2,3-epoxypropyl chloride, chloropropylene oxide
PubChem CID 7835
ChEBI CHEBI:37144
IUPAC Name 2-(chloromethyl)oxirane
SMILES C1C(O1)CCl

Specifications

Melting Point -57.0°C
Density 1.1800g/mL
Boiling Point 115.0°C to 117.0°C
Flash Point 28°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4365 to 1.4385
Quantity 2.5 L
Beilstein 17,6
Fieser 01,355; 04,222; 05,290
Merck Index 15,3666
Specific Gravity 1.18
Solubility Information Solubility in water: 60g/l (10°C). Other solubilities: misclible with alcohol, ether, chloroform,, trichloroethylene, carbon tetrachloride, acetone, and benzene
Viscosity 1.12 mPa.s (20°C)
Formula Weight 92.52
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Epichlorohydrin
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Causes severe skin burns and eye damage.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with
EINECSNumber 203-439-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.