Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epidermal Growth Factor-like Protein 7 Monoclonal Antibody (2H2), QED Bioscience™
SDP

Catalog No. 502278288
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8288 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8288 Supplier Leinco Technologies Inc Supplier No. 90013100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Epidermal Growth Factor-like Protein 7 Monoclonal antibody specifically detects Epidermal Growth Factor-like Protein 7 in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot

Specifications

Antigen Epidermal Growth Factor-like Protein 7
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2H2
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris with 0.1M glycine, 2% sucrose and no preservative
Gene EGFL7
Gene Accession No. Q9UHF1
Gene Alias Cbl20; EGF like domain multiple 7; EGFL7; EGF-like domain 7; EGF-like domain-containing protein 7; EGF-like protein 7; EGF-like-domain, multiple 7; Epidermal growth factor-like protein 7; Estrogen-regulated protein CBL20 20.4kD; Estrogen-regulated protein CBL20, 20.4kD; Megf7; multiple EGF-like domain protein 7; multiple EGF-like domains protein 7; multiple epidermal growth factor-like domain protein 7; multiple epidermal growth factor-like domains protein 7; NEU1; NEU1 protein; NOTCH4-like protein; RP11-251M1.2; UNQ187/PRO1449; Vascular endothelial statin; vascular endothelial-statin; VE-STATIN; Zneu1
Host Species Mouse
Immunogen His-tagged EGFL-7 recombinant protein. Accession no. Q9UHF1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51162
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.