Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epidermal Growth Factor Receptor/EGFR Ab-15 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS665P Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS665P 1 mL 200μg/mL
MS665P0 100 μL 200μg/mL
MS665P1 500 μL 200μg/mL
MS665P1ABX 100 μL 1mg/mL
MS665PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS665P Supplier Epredia Supplier No. MS665P

Ensure accurate, reproducible results in western blotting and immunoprecipitation experiments.

EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.

Host Species: Mouse

Clone: H9B4

Isotype: IgG1

Species Reactivity: Human. Does not react with mouse and rat. Others not known.

Epitope: Cytoplasmic domain

Immunogen: Purified EGFR from A431 cells

Molecular Weight: 170 kDa (wild type) and 145 kDa (vIII variant)

Positive Control: A431 cells

Cellular Localization: Membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Epidermal Growth Factor Receptor/EGFR Ab-15
Applications Immunoprecipitation
Classification Monoclonal
Clone H9B4
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Purified EGFR from A431 cells
Quantity 1 mL
Regulatory Status RUO
Research Discipline EGFR Family
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.