Learn More
Epidermal Growth Factor Receptor (EGFR) Rabbit Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Rabbit
Clone: EP38Y
Species Reactivity: Human. Others not tested.
Ig Isotype: Rabbit IgG
Epitope: Not determined
Immunogen: A synthetic phospho-peptide corresponding to residues surrounding Tyr1068 of human EGFR
Molecular Weight: 175 kDa
Positive Control: Squamous carcinoma
Cellular Localization: Cell membrane and occasional cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | EGFR |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Clone | EP38Y |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic phospho-peptide corresponding to residues surrounding Tyr1068 of human EGFR |
| Quantity | 12 mL |
| Primary or Secondary | Primary |
| Form | Ready-to-Use |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.