Description
Functions:
- Epidural anesthesia using the loss-of-resistance and the hanging-drop technique
- Spinal anesthesia with realistic resistance of the dura and arachnoid mater with or without a cannula
- Fluid-filled spinal canal with realistic outflow rate thanks to the ability to adjust the excess pressure
- Epidural catheter can be inserted into the epidural space
- Closed water system
- Easy to clean
- Suitable for both medical training and for specialist medical training.
Includes:
- Base body
- Vertebral bar
- LOR insert (loss-of-resistance)
- Skin
- Spinal canal with connector
- Water collection container
- 10 cm tube
- 2 x 100 mL injections with cap
- Spacer ring for injection
- Extension tube for the hanging-drop technique
- Carry bag
Specifications
Specifications
| Product Type | Epidural and Spinal Injection Trainer |
| Includes | Base body, Vertebral bar, LOR insert, Skin, Spinal canal with connector, Water collection container, 10 cm tube, 2 x 100 mL injections with cap, Spacer ring for injection, Extension tube for the hanging-drop technique, Carry bag |