Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Epigallocatechin, PROMO, Thermo Scientific Chemicals

Catalog No. J61630MAPM
Click to view available options
Quantity:
10 mg

Chemical Identifiers

CAS 970-74-1
Molecular Formula C15H14O7
Molecular Weight (g/mol) 306.27 g/mol
MDL Number MFCD00075939
InChI Key XMOCLSLCDHWDHP-IUODEOHRSA-N
Synonym --epigallocatechin, epigallocatechin, epigallocatechol, l-epigallocatechin, epi-gallocatechin, antiscurvy factor c2, --epigallocatechol, l-epigallocatechol, --3,3',4',5,5',7-flavanhexol, 3,3',4',5,5',7-flavanhexol
PubChem CID 72277
ChEBI CHEBI:42255
IUPAC Name (2R,3R)-2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C(=C3)O)O)O)O

Specifications

Color Pink
Quantity 10 mg
Physical Form Powder
Chemical Name or Material (-)-Epigallocatechin
RTECSNumber KB100000
TSCA No
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.