Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epimorphin/Syntaxin 2 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119338IR
Description
Epimorphin/Syntaxin 2 Polyclonal antibody specifically detects Epimorphin/Syntaxin 2 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Hamster, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Epimorphin/Syntaxin 2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
EPIMepimorphin, EPM, STX2AMGC51014, STX2Bsyntaxin-2, STX2CEpimorphin, syntaxin 2 | |
Synthetic peptide derived from sequence near the amino-terminus of rat Syntaxin 2, conjugated to KLH | |
0.1 mL | |
Neuroscience | |
2054 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Canine, Chicken | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction