Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epiregulin Antibody (6H12), PerCP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP221992PCP
Description
Epiregulin Monoclonal antibody specifically detects Epiregulin in Mouse samples. It is validated for ELISA, NeutralizationSpecifications
Epiregulin | |
Monoclonal | |
PerCP | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA, Neutralization | |
6H12 | |
ELISA, Block/Neutralize | |
epiregulin, ER, proepiregulin | |
Mouse recombinant Epiregulin | |
0.1 mL | |
Angiogenesis, Biologically Active Proteins | |
2069 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction